Learn More
Medchemexpress LLC Eya2 phosphatase inhibitor | 352336-36-8 | 99.4% | 341.36 g/mol | C17H12FN3O2S | 5 MG
SDP
Catalog No. 5000262825
Encompass_Preferred
MLS000544460 is a research-grade small molecule that selectively and reversibly inhibits the Eya2 phosphatase. It reduces Eya2-mediated cell migration and has reported anti-cancer activity. The compound is supplied as a powder and as DMSO solutions, is characterized by high purity, and is intended for laboratory research use only.
  • Selective, reversible inhibition of Eya2 phosphatase.
  • Reported Kd ≈ 2.0 μM and IC50 ≈ 4 μM for target engagement.
  • High purity (~99.4%) suitable for biochemical and cell-based assays.
  • Available as powder and as ready-to-use DMSO solutions.
  • Stable when stored under recommended conditions for research reagents.
  • Supplied with datasheet, certificate of analysis, and safety data sheet.
Catalog No. 50-002-62825 Supplier Medchemexpress LLC Supplier No. HY1335115MG
MLS000544460 is a research-grade small molecule that selectively and reversibly inhibits the Eya2 phosphatase. It reduces Eya2-mediated cell migration and has reported anti-cancer activity. The compound is supplied as a powder and as DMSO solutions, is characterized by high purity, and is intended for laboratory research use only.
  • Selective, reversible inhibition of Eya2 phosphatase.
  • Reported Kd ≈ 2.0 μM and IC50 ≈ 4 μM for target engagement.
  • High purity (~99.4%) suitable for biochemical and cell-based assays.
  • Available as powder and as ready-to-use DMSO solutions.
  • Stable when stored under recommended conditions for research reagents.
  • Supplied with datasheet, certificate of analysis, and safety data sheet.