Learn More
Aviva Systems Biology Corp Eya4 Antibody - N-terminal region (ARP39975_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39975_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Eya4. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N-terminal region of Eya4. Alternate names: B130023L16RikApplicationWB. Species reactivity: Dog, Guinea Pig, Human, Mouse, Rabbit, Zebrafish
Catalog No. 50-284-843