Learn More
Fine Science Tools Graefe Forceps, Stainless Steel, Curved, 10 cm
SDP
Catalog No. NC9643004
Encompass
Tip Shape: Curved/ Tips: Serrated/ Tip Dimensions: 0.8 x 0.7mm/ Alloy / Material: Stainless Steel/ Length: 10cm
Catalog No. NC9643004 Supplier Fine Science Tools Supplier No. 1105110
Tip Shape: Curved/ Tips: Serrated/ Tip Dimensions: 0.8 x 0.7mm/ Alloy / Material: Stainless Steel/ Length: 10cm