Learn More
Fine Science Tools Graefe Forceps, Stainless Steel, Straight, 10 cm
SDP
Catalog No. NC9486223
Encompass
Tip Shape: Straight/ Tips: Serrated/ Tip Dimensions: 0.8 x 0.7mm/ Alloy / Material: Stainless Steel/ Length: 10cm
Catalog No. NC9486223 Supplier Fine Science Tools Supplier No. 1105010
Tip Shape: Straight/ Tips: Serrated/ Tip Dimensions: 0.8 x 0.7mm/ Alloy / Material: Stainless Steel/ Length: 10cm