Learn More
Sklar Instruments Forceps, Surgi-OR Eye Dressing Forceps, Sklar in., Straight, Serrated, Stainless Steel, EA.
SDP
Catalog No. 501126555 Shop All Sklar Instruments Products
Encompass_Preferred
Forceps, Surgi-OR Eye Dressing Forceps, Sklar, 6 in., Straight, Serrated, Stainless Steel, EA. Surgi-OR Eye Dressing Forceps are thumb forceps used for holding dressing materials such as cotton and gauze during surgical procedures, changing dressings, or packing wounds. Hand-crafted in Pakistan, Surgi-OR mid-grade instruments are forged from the highest quality Japanese steel and provide reliable instrumentation for the operating room and other surgical centers. Surgi-OR instruments are a cost-effective line of instruments that must meet Sklar's quality standards. This product is straight with serrated tips and a length of 6 inches. Lifetime warranty against all manufacturer defects.
Catalog No. 50-112-6555 Supplier Sklar Instruments Supplier No. 95731
Forceps, Surgi-OR Eye Dressing Forceps, Sklar, 6 in., Straight, Serrated, Stainless Steel, EA. Surgi-OR Eye Dressing Forceps are thumb forceps used for holding dressing materials such as cotton and gauze during surgical procedures, changing dressings, or packing wounds. Hand-crafted in Pakistan, Surgi-OR mid-grade instruments are forged from the highest quality Japanese steel and provide reliable instrumentation for the operating room and other surgical centers. Surgi-OR instruments are a cost-effective line of instruments that must meet Sklar's quality standards. This product is straight with serrated tips and a length of 6 inches. Lifetime warranty against all manufacturer defects.