Learn More
Fine Science Tools EYE DRESSING FORCEPS/SER
SDP
Catalog No. NC9483144
Encompass
Tip Shape: Straight/ Tips: Serrated/ Tip Dimensions: 0.5 x 0.5mm/ Alloy / Material: Stainless Steel/ Length: 10cm
Catalog No. NC9483144 Supplier Fine Science Tools Supplier No. 1115010
Tip Shape: Straight/ Tips: Serrated/ Tip Dimensions: 0.5 x 0.5mm/ Alloy / Material: Stainless Steel/ Length: 10cm