Learn More
Grainger Eye End Fitting 316 SS 1/16 in Rope dia.
Catalog No. 502765655
Encompass_Preferred
The Loos Eye End Fitting is a 1/16" compatible eye end fitting made of carbon steel with a zinc finish. It has a maximum pressure rating of 3000 psi and temperature rating of 300°F. This fitting connects to 1/16" OD rigid metal tubing and has a hex head for easy tightening. It can be used for instrumentation, automation, and other low pressure hydraulic applications. The zinc finish provides corrosion resistance. This is an economical fitting for creating a swivel connection point on small tubing runs.
Catalog No. 50-276-5655 Supplier Grainger Supplier No. 16X678
The Loos Eye End Fitting is a 1/16" compatible eye end fitting made of carbon steel with a zinc finish. It has a maximum pressure rating of 3000 psi and temperature rating of 300°F. This fitting connects to 1/16" OD rigid metal tubing and has a hex head for easy tightening. It can be used for instrumentation, automation, and other low pressure hydraulic applications. The zinc finish provides corrosion resistance. This is an economical fitting for creating a swivel connection point on small tubing runs.