Learn More
Grainger Eye End Fitting 316 SS 1/4 in Rope dia.
Catalog No. 502767220
Encompass_Preferred
The Grainger 316 stainless steel eye fittings for wire rope are designed for use with wire rope or cable to form an eye for attaching fittings. They are made from 316 stainless steel for strength and corrosion resistance. These fittings are available in multiple sizes to accommodate wire rope diameters from 1/8" to 1" and feature a threaded opening to accept various threaded studs. The working load limit ranges from 1,260 lbs to 23,000 lbs depending on the size. These fittings meet military, federal, and other specifications and are manufactured in the USA. They provide a secure method of forming an eye on the end of a wire rope for lifting and rigging applications.
Catalog No. 50-276-7220 Supplier Grainger Supplier No. 16X684
The Grainger 316 stainless steel eye fittings for wire rope are designed for use with wire rope or cable to form an eye for attaching fittings. They are made from 316 stainless steel for strength and corrosion resistance. These fittings are available in multiple sizes to accommodate wire rope diameters from 1/8" to 1" and feature a threaded opening to accept various threaded studs. The working load limit ranges from 1,260 lbs to 23,000 lbs depending on the size. These fittings meet military, federal, and other specifications and are manufactured in the USA. They provide a secure method of forming an eye on the end of a wire rope for lifting and rigging applications.