Learn More
Grainger Eye End Fitting 316 SS 1/8 in Rope dia.
Catalog No. 502765682
Encompass_Preferred
The Loos & Co. 1/8" Eye End Fitting is a forged steel fitting with a zinc finish and an eye end connection. It has a pressure rating of 3000 psi and temperature rating of 300°F. This fitting has a thread size of 1/8" NPT and is made in the USA. It can be used for hydraulic and pneumatic applications. The eye end has a 0.28" hole diameter. This is an economical fitting for use in industrial environments.
Catalog No. 50-276-5682 Supplier Grainger Supplier No. 16X680
The Loos & Co. 1/8" Eye End Fitting is a forged steel fitting with a zinc finish and an eye end connection. It has a pressure rating of 3000 psi and temperature rating of 300°F. This fitting has a thread size of 1/8" NPT and is made in the USA. It can be used for hydraulic and pneumatic applications. The eye end has a 0.28" hole diameter. This is an economical fitting for use in industrial environments.