Learn More
Grainger Eye End Fitting 316 SS 3/16 in Rope dia.
Catalog No. 502765640
Encompass_Preferred
The Grainger 16X682 is a 1/2 HP, 115V, 1725 RPM, 56C Frame, Open Drip Proof, Single Phase, Ball Bearing, Electric Motor. It has a 48Y frame with a 1.15 service factor and features a cast iron body with an aluminum end bell. This motor is thermally protected and suitable for continuous duty applications. The 16X682 has a C-face mounting style and comes with standard leads.
Catalog No. 50-276-5640 Supplier Grainger Supplier No. 16X682
The Grainger 16X682 is a 1/2 HP, 115V, 1725 RPM, 56C Frame, Open Drip Proof, Single Phase, Ball Bearing, Electric Motor. It has a 48Y frame with a 1.15 service factor and features a cast iron body with an aluminum end bell. This motor is thermally protected and suitable for continuous duty applications. The 16X682 has a C-face mounting style and comes with standard leads.