Learn More
Grainger Eye End Fitting 316 SS 5/32 in Rope dia.
Catalog No. 502767139
Encompass_Preferred
The LOOS Eye End Fitting is a 5/32" compatible eye end fitting made of carbon steel with a zinc finish. It has a working pressure of 3,000 psi and temperature range of -20 to 300 degrees Fahrenheit. This fitting has a thread size of 5/32-24 UNF-2A and is designed for use with 5/32" OD rigid metal tubing. The overall dimensions are 15/32" diameter x 27/64" length. This eye end fitting provides a secure 90 degree bend in hydraulic and pneumatic applications.
Catalog No. 50-276-7139 Supplier Grainger Supplier No. 16X681
The LOOS Eye End Fitting is a 5/32" compatible eye end fitting made of carbon steel with a zinc finish. It has a working pressure of 3,000 psi and temperature range of -20 to 300 degrees Fahrenheit. This fitting has a thread size of 5/32-24 UNF-2A and is designed for use with 5/32" OD rigid metal tubing. The overall dimensions are 15/32" diameter x 27/64" length. This eye end fitting provides a secure 90 degree bend in hydraulic and pneumatic applications.