Learn More
Grainger Eye End Fitting 316 SS 7/32 in Rope dia.
Catalog No. 502768010
Encompass_Preferred
The Loos Eye End Fitting is made of 316 stainless steel and designed for use with 7/32 inch diameter rope. It has a working load limit of 550 lbs and meets military specification MIL-S-15719F. The fitting has a threaded stud for securing to structures. Overall length is 1-3/4 inches and it weighs approximately 1 oz. This high quality stainless steel fitting provides a strong and durable end termination for 7/32 inch diameter rope.
Catalog No. 50-276-8010 Supplier Grainger Supplier No. 16X683
The Loos Eye End Fitting is made of 316 stainless steel and designed for use with 7/32 inch diameter rope. It has a working load limit of 550 lbs and meets military specification MIL-S-15719F. The fitting has a threaded stud for securing to structures. Overall length is 1-3/4 inches and it weighs approximately 1 oz. This high quality stainless steel fitting provides a strong and durable end termination for 7/32 inch diameter rope.