Learn More
Grainger Eye End Fitting 316 SS 9/32 in Rope dia.
Catalog No. 502765641
Encompass_Preferred
The LOOS Eye End Fitting is a 9/32" compatible eye end fitting made of carbon steel with a zinc finish. It has a working pressure of 3,000 psi and temperature range of -20 to 300 degrees Fahrenheit. This fitting connects to 9/32" OD rigid metal tubing and has a hex head for easy tightening. It can be used for instrumentation, automation, and other industrial applications using metal tubing. The LOOS eye end fitting is RoHS compliant.
Catalog No. 50-276-5641 Supplier Grainger Supplier No. 16X685
The LOOS Eye End Fitting is a 9/32" compatible eye end fitting made of carbon steel with a zinc finish. It has a working pressure of 3,000 psi and temperature range of -20 to 300 degrees Fahrenheit. This fitting connects to 9/32" OD rigid metal tubing and has a hex head for easy tightening. It can be used for instrumentation, automation, and other industrial applications using metal tubing. The LOOS eye end fitting is RoHS compliant.