Learn More
Grainger Eye End Fitting SS 1/4 in Rope dia.
Catalog No. 502767222
Encompass_Preferred
The Loos & Co. 1/4" Eye End Fitting is a forged steel fitting with a zinc finish and an eye end connection. It has a pressure rating of 3000 psi and temperature rating of 300°F. This fitting has a thread size of 1/4" NPT and is made in the USA. It can be used for hydraulic and pneumatic applications. The overall dimensions are 1-1/2" length x 1" height x 1-1/8" width.
Catalog No. 50-276-7222 Supplier Grainger Supplier No. 16X674
The Loos & Co. 1/4" Eye End Fitting is a forged steel fitting with a zinc finish and an eye end connection. It has a pressure rating of 3000 psi and temperature rating of 300°F. This fitting has a thread size of 1/4" NPT and is made in the USA. It can be used for hydraulic and pneumatic applications. The overall dimensions are 1-1/2" length x 1" height x 1-1/8" width.