Learn More
Grainger Eye End Fitting SS 3/16 in Rope dia.
Catalog No. 502768193
Encompass_Preferred
The Loos & Co. 3/16" Stainless Steel Eye End Fitting is made from 316 stainless steel for corrosion resistance. It has a working load limit of 1,400 lbs and is designed for use with 3/16" diameter wire rope. The opening measures 5/8" diameter. This eye end fitting creates a permanent loop and attaches with two ferrule crimps. It meets military specification MIL-T-781. The overall length is 1-3/4". This eye end fitting is ideal for creating an endless wire rope sling.
Catalog No. 50-276-8193 Supplier Grainger Supplier No. 16X672
The Loos & Co. 3/16" Stainless Steel Eye End Fitting is made from 316 stainless steel for corrosion resistance. It has a working load limit of 1,400 lbs and is designed for use with 3/16" diameter wire rope. The opening measures 5/8" diameter. This eye end fitting creates a permanent loop and attaches with two ferrule crimps. It meets military specification MIL-T-781. The overall length is 1-3/4". This eye end fitting is ideal for creating an endless wire rope sling.