Learn More
Grainger Eye End Fitting SS 5/16 in Rope dia.
Catalog No. 502765069
Encompass_Preferred
The Loos Eye End Fitting is a 5/16 inch eye end fitting made of carbon steel with a zinc finish. It has a working pressure of 3,000 psi and temperature range of -20 to 300 degrees Fahrenheit. This fitting creates a loop and attaches to the end of a hose or tube. It can be used for hydraulic and pneumatic applications. The carbon steel material provides strength while the zinc coating protects against corrosion. This is an economic fitting for industrial applications requiring an eye end connection.
Catalog No. 50-276-5069 Supplier Grainger Supplier No. 16X676
The Loos Eye End Fitting is a 5/16 inch eye end fitting made of carbon steel with a zinc finish. It has a working pressure of 3,000 psi and temperature range of -20 to 300 degrees Fahrenheit. This fitting creates a loop and attaches to the end of a hose or tube. It can be used for hydraulic and pneumatic applications. The carbon steel material provides strength while the zinc coating protects against corrosion. This is an economic fitting for industrial applications requiring an eye end connection.