Learn More
Grainger Haws Axion MSR Eye/Face Wash, Semi-Recessed, Stainless Steel, Wall-Mounted, 1 ea
Catalog No. 502791770
Encompass_Preferred
The semi-recessed wall mount plumbed eye/face wash provides reliable emergency rinsing for both the eyes and face. Designed for accessibility, this unit features a durable 18 gauge, Type 304 stainless steel cabinet and a polished chrome-plated brass pull-down valve for easy activation. It utilizes proprietary technology to deliver an inverted directional laminar flow, meeting high-performance standards.
  • Features AXION® MSR eye/face wash head with inverted directional laminar flow
  • Constructed from 18 gauge, Type 304 stainless steel with a front-access maintenance panel
  • Compliant with ANSI Z358.1 standards
  • Includes brass pipe and fittings, universal identification sign, and inspection tag
  • Designed for wheelchair accessibility
  • Operates at 30 psi to 90 psi flowing pressure
  • Inlet supply is male 1/2 in. IPS
Catalog No. 50-279-1770 Supplier Grainger Supplier No. 16D395
The semi-recessed wall mount plumbed eye/face wash provides reliable emergency rinsing for both the eyes and face. Designed for accessibility, this unit features a durable 18 gauge, Type 304 stainless steel cabinet and a polished chrome-plated brass pull-down valve for easy activation. It utilizes proprietary technology to deliver an inverted directional laminar flow, meeting high-performance standards.
  • Features AXION® MSR eye/face wash head with inverted directional laminar flow
  • Constructed from 18 gauge, Type 304 stainless steel with a front-access maintenance panel
  • Compliant with ANSI Z358.1 standards
  • Includes brass pipe and fittings, universal identification sign, and inspection tag
  • Designed for wheelchair accessibility
  • Operates at 30 psi to 90 psi flowing pressure
  • Inlet supply is male 1/2 in. IPS