Learn More
Grainger Haws Eye/Face Wash Pedestal, Stainless Steel, Floor Mounted, 11 in. Bowl, ANSI Z358.1 Compliant
Catalog No. 502790550
Encompass_Preferred
The pedestal-mounted eye/face wash system is designed for emergency use in environments exposed to corrosive materials, providing immediate flushing to protect the eyes and face. The unit features a specialized wash head that utilizes inverted directional laminar flow to sweep contaminants away from the nasal cavity. Constructed from high-quality stainless steel, this durable system includes an integrated water strainer to maintain optimal performance and prevent debris interference.
  • Features AXION™ MSR eye/face wash head with inverted directional laminar flow and zero vertical velocity
  • Constructed with Type 304 stainless steel pipe, fittings, and an 11 in. round bowl
  • Equipped with a Type 316 stainless steel ball valve for reliable corrosion resistance
  • Includes a 50 x 50 mesh in-line water strainer for debris prevention
  • Certified by CSA to meet the ANSI/ISEA Z358.1 standard for emergen
Catalog No. 50-279-0550 Supplier Grainger Supplier No. 16D374
The pedestal-mounted eye/face wash system is designed for emergency use in environments exposed to corrosive materials, providing immediate flushing to protect the eyes and face. The unit features a specialized wash head that utilizes inverted directional laminar flow to sweep contaminants away from the nasal cavity. Constructed from high-quality stainless steel, this durable system includes an integrated water strainer to maintain optimal performance and prevent debris interference.
  • Features AXION™ MSR eye/face wash head with inverted directional laminar flow and zero vertical velocity
  • Constructed with Type 304 stainless steel pipe, fittings, and an 11 in. round bowl
  • Equipped with a Type 316 stainless steel ball valve for reliable corrosion resistance
  • Includes a 50 x 50 mesh in-line water strainer for debris prevention
  • Certified by CSA to meet the ANSI/ISEA Z358.1 standard for emergen