Learn More
Grainger Haws Axion MSR Eye/Face Wash, Wall Mount, Stainless Steel, 1 ea
Catalog No. 502791886
Encompass_Preferred
The AXION® MSR corrosion-resistant wall-mount eye/face wash provides an emergency solution designed to sweep contaminants away from the nasal cavity using an inverted directional laminar flow. This unit is pre-built and fully pressure-tested to reduce installation time. It is certified by CSA to meet the ANSI/ISEA Z358.1 standard for emergency eyewash and shower equipment.
  • Inverted flow design with Zero Vertical Velocity™ stream engineering directs contaminants away from the nasal cavity
  • 16-gauge Type 316 stainless steel ball valve with stainless steel ball and stem provides corrosion resistance
  • Type 304 stainless steel in-line 50 x 50 mesh water strainer prevents debris from reaching the wash head
  • Stainless steel dust cover protects the bowl and wash head until use
  • Includes 20.3 cm x 27.3 cm (8 in. x 10-3/4 in.) universal emergency sign and maintenance test card
Catalog No. 50-279-1886 Supplier Grainger Supplier No. 16D398
The AXION® MSR corrosion-resistant wall-mount eye/face wash provides an emergency solution designed to sweep contaminants away from the nasal cavity using an inverted directional laminar flow. This unit is pre-built and fully pressure-tested to reduce installation time. It is certified by CSA to meet the ANSI/ISEA Z358.1 standard for emergency eyewash and shower equipment.
  • Inverted flow design with Zero Vertical Velocity™ stream engineering directs contaminants away from the nasal cavity
  • 16-gauge Type 316 stainless steel ball valve with stainless steel ball and stem provides corrosion resistance
  • Type 304 stainless steel in-line 50 x 50 mesh water strainer prevents debris from reaching the wash head
  • Stainless steel dust cover protects the bowl and wash head until use
  • Includes 20.3 cm x 27.3 cm (8 in. x 10-3/4 in.) universal emergency sign and maintenance test card