Learn More
Med Vet International EYE Hook Set , J1369, Each
Catalog No. 5000584141
Encompass_Preferred
Blunt, atraumatic hooks held together by a heavy duty rubber band. The created tension holds them in place in both eye sockets when traction is exerted keeping the head straight and aligned in position. Stainless steel hooks. Pair.
Catalog No. 50-005-84141 Supplier Med Vet International Supplier No. JOR1369
Blunt, atraumatic hooks held together by a heavy duty rubber band. The created tension holds them in place in both eye sockets when traction is exerted keeping the head straight and aligned in position. Stainless steel hooks. Pair.