Learn More
Grainger Acorn Eye Sensor Assembly, 9 VDC, Programmable, 1 ea.
Catalog No. 502785177
Encompass_Preferred
This eye sensor assembly is designed for use with stainless steel wash fountains. It operates at 9 VDC and includes integrated wiring and plug clips for a complete installation. The unit features a programmable piezo pushbutton and solenoid assembly to facilitate efficient sensor-activated operation.
  • Operates on 9 VDC power
  • Designed for use with stainless steel wash fountains
  • Includes wiring and plug clips
  • Features programmable piezo pushbutton and solenoid components
Catalog No. 50-278-5177 Supplier Grainger Supplier No. 19C771
This eye sensor assembly is designed for use with stainless steel wash fountains. It operates at 9 VDC and includes integrated wiring and plug clips for a complete installation. The unit features a programmable piezo pushbutton and solenoid assembly to facilitate efficient sensor-activated operation.
  • Operates on 9 VDC power
  • Designed for use with stainless steel wash fountains
  • Includes wiring and plug clips
  • Features programmable piezo pushbutton and solenoid components