Learn More
Med Vet International McKesson Eye Wash Solution, 1-ounce Squeeze Bottle, MCK19828, Each
Catalog No. 503043180
Encompass_Preferred
Eye Wash Solution McKesson Active ingredient: 98.3% Purified Water Inactive ingredients: boric acid, sodium borate, sodium chloride 1 oz. Squeeze Bottle 98.3% purified water Cleanses the eye and helps relieve irritation or burning by removing loose foreign material Screw cap protected by a plastic tamper-evident seal The rate of flow is affected by the pressure applied to the bottle Sterile
Catalog No. 50-304-3180 Supplier Med Vet International Supplier No. 1188884EA
Eye Wash Solution McKesson Active ingredient: 98.3% Purified Water Inactive ingredients: boric acid, sodium borate, sodium chloride 1 oz. Squeeze Bottle 98.3% purified water Cleanses the eye and helps relieve irritation or burning by removing loose foreign material Screw cap protected by a plastic tamper-evident seal The rate of flow is affected by the pressure applied to the bottle Sterile