Learn More
Grainger GUARDIAN EQUIPMENT Eyewash Preservative, 8 oz
Catalog No. NC2182883
Encompass_Preferred
Guardian Equipment Eyewash Preservative is an 8 oz bottle of sterile solution designed to preserve eyewash stations. It helps prevent bacterial growth in eyewash stations for up to 6 months. This FDA-approved solution contains stabilized sodium hypochlorite to inhibit bacteria, fungi, and algae. It is compatible with most materials and won't corrode eyewash station nozzles. Easy to use - just pour into the eyewash reservoir. This product helps maintain eyewash station cleanliness and readiness for emergency use.
This item is not returnable. View return policy
Catalog No. NC2182883 Supplier Grainger Supplier No. 2LVL5
Guardian Equipment Eyewash Preservative is an 8 oz bottle of sterile solution designed to preserve eyewash stations. It helps prevent bacterial growth in eyewash stations for up to 6 months. This FDA-approved solution contains stabilized sodium hypochlorite to inhibit bacteria, fungi, and algae. It is compatible with most materials and won't corrode eyewash station nozzles. Easy to use - just pour into the eyewash reservoir. This product helps maintain eyewash station cleanliness and readiness for emergency use.