Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eye Wonder Book Series
SCI_ED

Catalog No. S79147
Change view
Click to view available options
Product Type:
Ocean
Space
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
S79147 Space
S79146 Ocean
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S79147 Supplier Penguin Random House LLC Supplier No. 9780789478542

missing translation for 'validMainframeMsg'

Open students' eyes to a world of discovery

  • Reference series specially developed for young students ready to read about the natural world
  • Written in text that is clear, understandable, informative
  • Photography depicts subjects in their natural settings
  • Hardcover; 48 pages; Grades K-3

Bugs
Explores the world of insects from dung beetles to dragonflies and more. 2002.

Human Body
Brief explanation of the human body; also includes some facts about sleep. 2003.

Dinosaur
Introduces students to many types of dinosaurs such as the Tyrannosaurus Rex, the Velociraptor, and the Hadrosaur. 2001.

Earth
Presents general information about the earth with high-quality images. 2002.

Human Body
Brief explanation of the human body; also includes some facts about sleep. 2003.

Ocean
Describes coral reefs, kelp forests, and other underwater worlds, animals such as whales, dolphins, sharks, and more. 2001.

Space
Guide to outer space and astronomy that includes information about exploration, solar system, planets, stars, and galaxies. 2001.

Rocks and Minerals
Teaches geology topics to young students such as the characteristics of rocks and minerals. 2003.

Volcano
Answers questions like How are volcanoes formed? and How hot is molten lava? Also presents information about famous volcanic eruptions. 2003.

Specifications

Grade K to 8
Format Book
Product Type Space
For Use With (Application) Guide to outer space and astronomy that includes information about exploration, solar system, planets, stars and galaxies
Subject Physics
Author Simon Holland
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.