Learn More
MSC EYEGLASS FRME/MNT W/CASE
Catalog No. 5000769518
Encompass
This eyeglass frame and mount is designed for use with 3M™ Full Facepiece Respirators 7000 Series, allowing the wearer to use corrective lenses comfortably.
  • Permits the wearer to use corrective lenses
  • Allows for comfortable position adjustment
  • For industrial/occupational use only
  • Compatible with Full Facepiece Respirators 7000 Series
Catalog No. 50-007-69518 Supplier MSC Supplier No. 57256885
This eyeglass frame and mount is designed for use with 3M™ Full Facepiece Respirators 7000 Series, allowing the wearer to use corrective lenses comfortably.
  • Permits the wearer to use corrective lenses
  • Allows for comfortable position adjustment
  • For industrial/occupational use only
  • Compatible with Full Facepiece Respirators 7000 Series