Learn More
Roboz Surgical EYEGLASSES, 2.5X MAGNIFICATION
SDP
Catalog No. NC2250820
Encompass
Loupes on Sports Eyeglasses, 2.5X magnification, 420mm working distance.
This item is not returnable. View return policy
Catalog No. NC2250820 Supplier Roboz Surgical Supplier No. ML613
Loupes on Sports Eyeglasses, 2.5X magnification, 420mm working distance.