Learn More
Electron Microscopy Sciences Eyelash Manipulator
SDP
Catalog No. NC1083208
Encompass_Preferred
For direct manipulation of sections and small tissue samples this tool is perfect for any occasion. May be used during ultramicrotomy to carefully tease sections that are in the Diamond Knife boat. The Eyelash comes on a Sable Brush handle made from the highest quality wood. These Eyelashes may be cleaned and sterilized with alcohol and a tissue.
Catalog No. NC1083208 Supplier Electron Microscopy Sciences Supplier No. 71182
For direct manipulation of sections and small tissue samples this tool is perfect for any occasion. May be used during ultramicrotomy to carefully tease sections that are in the Diamond Knife boat. The Eyelash comes on a Sable Brush handle made from the highest quality wood. These Eyelashes may be cleaned and sterilized with alcohol and a tissue.