Learn More
Meiji Techno America Meiji Techno America Eyepiece micrometer, 5mm divided into 100 units, 25mm diameter
Catalog No. NC1097493
Encompass_Preferred
Eyepiece micrometer, 5mm divided into 100 units, 25mm diameter
Catalog No. NC1097493 Supplier Meiji Techno America Supplier No. MA509
Eyepiece micrometer, 5mm divided into 100 units, 25mm diameter