Learn More
Cole-Parmer Meiji Techno MA506 Microscope Eyepiece Reticle, 10 mm/0.1 mm scale
Catalog No. 50631639 Shop All Cole-Parmer Products
Encompass
Use these micrometers to make quick and accurate measurements of your specimen. Eyepiece reticles are available with a crosshair scale or a 10 mm scale in 0.1 mm divisions; stage micrometers are available with a 1 mm scale in 0.01 mm divisions or with a 0.04" scale in 0.001" divisions. Use stage micrometers with any microscope.
Catalog No. 50-631-639 Supplier Cole-Parmer Supplier No. 4840486
Use these micrometers to make quick and accurate measurements of your specimen. Eyepiece reticles are available with a crosshair scale or a 10 mm scale in 0.1 mm divisions; stage micrometers are available with a 1 mm scale in 0.01 mm divisions or with a 0.04" scale in 0.001" divisions. Use stage micrometers with any microscope.