Learn More
Cole-Parmer Meiji Techno MA503 Microscope eyepieces; magnification, 15x; field of view, 15.4 mm
Catalog No. 50631637 Shop All Cole-Parmer Products
Encompass
These paired widefield eyepieces give you bright, sharp images. Reticle mount included. Eyepiece dia is 25 mm ID x 34 mm OD.
Catalog No. 50-631-637 Supplier Cole-Parmer Supplier No. 4840260
These paired widefield eyepieces give you bright, sharp images. Reticle mount included. Eyepiece dia is 25 mm ID x 34 mm OD.