Learn More
Cole-Parmer Meiji Techno MA504 Microscope eyepieces; magnification, 20x; field of view, 11.5 mm
Catalog No. 50631638 Shop All Cole-Parmer Products
Encompass
These paired widefield eyepieces give you bright, sharp images. Reticle mount included. Eyepiece dia is 25 mm ID x 34 mm OD.
Catalog No. 50-631-638 Supplier Cole-Parmer Supplier No. 4840262
These paired widefield eyepieces give you bright, sharp images. Reticle mount included. Eyepiece dia is 25 mm ID x 34 mm OD.