Learn More
Med Vet International Jorvet Eyesaver Eye Patch, Double Sided, Left, X-Large, J0478L
Catalog No. 5000495362
Encompass_Preferred
JorVet EyeSaver eye patch, is the ultimate in eye care. It features the exclusive Equine Eye Patch.EyeSaver's Eye Patch that effectively closes the horse's eye for maximum pain relief while allowing for constant monitoring of the injury or problem. EyeSaver affords the eye the most complete rest possible, eliminating the need for surgical closure. Don't let minor injuries become major problems-order your EyeSavers now. You'll be glad you did. EyeSaver Kit contains: EyeSaver hood 3 tear pads 3 snap-on rings 25 gauze pads
Catalog No. 50-004-95362 Supplier Med Vet International Supplier No. JOR478L
JorVet EyeSaver eye patch, is the ultimate in eye care. It features the exclusive Equine Eye Patch.EyeSaver's Eye Patch that effectively closes the horse's eye for maximum pain relief while allowing for constant monitoring of the injury or problem. EyeSaver affords the eye the most complete rest possible, eliminating the need for surgical closure. Don't let minor injuries become major problems-order your EyeSavers now. You'll be glad you did. EyeSaver Kit contains: EyeSaver hood 3 tear pads 3 snap-on rings 25 gauze pads