Learn More
ORS Nasco EYESLIN EYE WASHBTL 8OZ 12/CS
Catalog No. 502551604
Encompass_Preferred
Honeywell part# 32-000445-0000. Honeywell Eyesaline Personal Eyewash Bottle Includes: Buffered Saline solution and superior to tap Water temper resistant seal instructions Capacity: 8 oz. 236.5mL Certifications/Compliance: FDA
Catalog No. 50-255-1604 Supplier ORS Nasco Supplier No. 203320004450000
Honeywell part# 32-000445-0000. Honeywell Eyesaline Personal Eyewash Bottle Includes: Buffered Saline solution and superior to tap Water temper resistant seal instructions Capacity: 8 oz. 236.5mL Certifications/Compliance: FDA