Learn More
ORS Nasco Personal Eye Wash Solution, 16 oz, Bottle
Catalog No. 502563474
Encompass_Preferred
Honeywell Part # 32-000454-0000-H5. Designed for flushing nuisance particles such as pollen, dust, sawdust, smoke as well as for immediate treatment. Bottles contain a buffered, saline solution that is sterile and superior to tap water for emergency eye care. Blow-fill-seal bottles with extended-flow nozzles are tamper resistant, yet easily removed in an emergency. Deliver drops or an irrigating stream of solution, depending on the squeeze pressure applied to the bottle.
Catalog No. 50-256-3474 Supplier ORS Nasco Supplier No. 203320004540000H5
Honeywell Part # 32-000454-0000-H5. Designed for flushing nuisance particles such as pollen, dust, sawdust, smoke as well as for immediate treatment. Bottles contain a buffered, saline solution that is sterile and superior to tap water for emergency eye care. Blow-fill-seal bottles with extended-flow nozzles are tamper resistant, yet easily removed in an emergency. Deliver drops or an irrigating stream of solution, depending on the squeeze pressure applied to the bottle.