Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mopec Eyewash Assembly, Hose Style
SDP

Catalog No. 22044650 Shop All Mopec Products
Change view
Click to view available options
Color:
Gray
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
22-044-650 Gray
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-044-650 Supplier Mopec Supplier No. OA111
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Hose-style eyewash assembly provides a flexible solution for emergency eye rinsing in laboratory and clinical environments. Handheld hose allows targeted flushing for quick response and supports workplace safety.

Construction

  • Hand-held
  • 3.2 gpm once activated
  • Single spray head
  • Squeeze handle to begin the flow of water
  • Release handle to stop the flow of water

Dimensions

  • 1 to 1/2 in. spray head diameter
  • 3/8 in. inlet pipe size

Materials

  • 8 ft. PVC hose
  • HDPE spray head

Utilities / Power

  • 300 PSI maximum water pressure

Specifications

Color Gray
Flow Rate 3.2 gpm
Material PVC
Mounting Surface
Product Type Eyewash Assembly
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.