Learn More
Water Saver Faucet Company EYEWASH/DRENCH HOSE UNT, DK MT
Catalog No. NC2212255
Encompass_Preferred
Eyewash/Drench Hose Unit, Deck Mounted, Ball Valve with Flag Handle, Vacuum Breaker
This item is not returnable. View return policy
Catalog No. NC2212255 Supplier Water Saver Faucet Company Supplier No. EW1028VB
Eyewash/Drench Hose Unit, Deck Mounted, Ball Valve with Flag Handle, Vacuum Breaker