Learn More
Grainger Haws Plumbed Eyewash Station, Wall Mount, Freeze Resistant, 1 ea
Catalog No. 502791916
Encompass_Preferred
This plumbed wall-mount eyewash station is specifically designed for use in environments prone to freezing temperatures. The unit features freeze-resistant valves and supply lines, which are installed in a heated space on the opposite side of a wall to ensure reliable operation and prevent line freezing. Each unit fully drains after use to eliminate standing water. It includes a stainless steel bowl and is activated via a push plate, providing effective eye coverage for emergency situations.
  • Designed for operation in freezing temperature conditions
  • Self-draining valve system prevents water from freezing in lines
  • Constructed with a 27.9 cm (11 in.) diameter stainless steel bowl
  • Equipped with anti-surge, soft-flo plastic spray heads with integral flow controls and dust covers
  • Meets ANSI Z358.1 requirements
  • Features a 1/2 in. NPT female inlet and a 1-1/4 in. NPT outlet
  • Includes
Catalog No. 50-279-1916 Supplier Grainger Supplier No. 16D390
This plumbed wall-mount eyewash station is specifically designed for use in environments prone to freezing temperatures. The unit features freeze-resistant valves and supply lines, which are installed in a heated space on the opposite side of a wall to ensure reliable operation and prevent line freezing. Each unit fully drains after use to eliminate standing water. It includes a stainless steel bowl and is activated via a push plate, providing effective eye coverage for emergency situations.
  • Designed for operation in freezing temperature conditions
  • Self-draining valve system prevents water from freezing in lines
  • Constructed with a 27.9 cm (11 in.) diameter stainless steel bowl
  • Equipped with anti-surge, soft-flo plastic spray heads with integral flow controls and dust covers
  • Meets ANSI Z358.1 requirements
  • Features a 1/2 in. NPT female inlet and a 1-1/4 in. NPT outlet
  • Includes