Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Haws™ Eyewash Head with Flow Control
SDP

Catalog No. 19039802 Shop All Haws Corporation Products
Change view
Click to view available options
For Use With (Equipment):
Eye and Face Wash
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
19-039-802 Eye and Face Wash
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-039-802 Supplier Haws™ Supplier No. SP11FC
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Assures a single, soft-and-steady stream of water for flowing water pressure ranging from 30 to 90 psi (2.1 to 6.2 bar).

  • Built-in 1.2 gpm (4.5L) automatic pressure compensation flow control
  • Flow control is easily accessible for cleaning
  • Soft-Flo ABS plastic
  • Anti-surge
  • Green
  • 1/4 in. IPS

Specifications

Color Green
For Use With (Equipment) Eye and Face Wash
Includes Built-in 1.2 gpm (4.5 L) Automatic Pressure Compensation Flow Control
Material ABS Plastic
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.