Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Haws™ Eyewash Head with Nozzle
SDP

Catalog No. 19811868 Shop All Haws Corporation Products
Change view
Click to view available options
For Use With (Equipment):
Portable Tank Eyewash Units
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
19-811-868 Portable Tank Eyewash Units
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-811-868 Supplier Haws™ Supplier No. SP12
  • $149.00
    $111.65 / Each of 1

    Save $37.35

    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For use with portable tank eyewash units.

  • Green ABS plastic
  • Anti-surge Soft-Flo head
  • Inserted nozzle in orifice
  • For use with portable tank eyewash units
  • 1/4 in. IPS.

Specifications

Color Green
For Use With (Equipment) Portable Tank Eyewash Units
Material ABS Plastic
Product Type Eyewash Head
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.