Learn More
Grainger EYEWASH REPL EYEWASH BTL
Catalog No. NC9322304
Encompass_Preferred
Replacement Bottle, with 32oz. eyesaline, ANSI Z358.1-1998, Durable plastic stations available with 16- or 32-oz. bottles, Bottles have twist-off tops and no seals expiration dated to guard against contamination, 1/PK
Catalog No. NC9322304 Supplier Grainger Supplier No. 3ARE1
Replacement Bottle, with 32oz. eyesaline, ANSI Z358.1-1998, Durable plastic stations available with 16- or 32-oz. bottles, Bottles have twist-off tops and no seals expiration dated to guard against contamination, 1/PK