Learn More
ORS Nasco EYEWASH SELF CONTAINED 15 MINS
Catalog No. 502551600
Encompass_Preferred
Honeywell part# 32-001000-0000. Honeywell Pure Flow 1000 Portable Emergency Eyewash Station Capacity: 892.6 oz. 26397.33mL Certifications/Compliance: ANSI Z358.1-2004 Depth: 17.25 in. 43.81cm Height: 30 in. 76.2 cm
Catalog No. 50-255-1600 Supplier ORS Nasco Supplier No. 203320010000000
Honeywell part# 32-001000-0000. Honeywell Pure Flow 1000 Portable Emergency Eyewash Station Capacity: 892.6 oz. 26397.33mL Certifications/Compliance: ANSI Z358.1-2004 Depth: 17.25 in. 43.81cm Height: 30 in. 76.2 cm