Learn More
Grainger HONEYWELL, Single-Use Eyewash Station 32OZ
Catalog No. NC1954899
Encompass_Preferred
The Honeywell Single-Use Eyewash Station is a portable, disposable eyewash solution designed for emergency eye flushing after exposure to dust, debris, chemicals or other eye irritants. It contains sterile saline solution in a sealed soft plastic bottle with an angled eyecup designed for hands-free rinsing. No batteries or activation required. Can provide 15 minutes of gentle flushing. Single use only. Compact size is easy to store anywhere eyewash access is needed. Provides ANSI-compliant eye flushing in locations lacking plumbed emergency stations.
Catalog No. NC1954899 Supplier Grainger Supplier No. 3ARD8
The Honeywell Single-Use Eyewash Station is a portable, disposable eyewash solution designed for emergency eye flushing after exposure to dust, debris, chemicals or other eye irritants. It contains sterile saline solution in a sealed soft plastic bottle with an angled eyecup designed for hands-free rinsing. No batteries or activation required. Can provide 15 minutes of gentle flushing. Single use only. Compact size is easy to store anywhere eyewash access is needed. Provides ANSI-compliant eye flushing in locations lacking plumbed emergency stations.