Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Notrax Eyewash Station Mat

Catalog No. 17004380 Shop All NoTrax Products
Change view
Click to view available options
Dimensions (L x W):
36 x 30 in.
36 x 42 in.
48 x 42 in.
48 x 54 in.
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
17-004-380 48 x 54 in.
17-001-098 36 x 42 in.
17-001-099 48 x 42 in.
17-000-033 36 x 30 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 17-004-380 Supplier Notrax Supplier No. 524S5448GN
Add to Cart
Add to Cart

Space in front of emergency response stations, including eyewash and showers, is often viewed as empty space. This emergency mat is highly visible due to its bright safety colors.

  • The open space in front of emergency response stations and equipment is often viewed as empty space that can be used for storage.
  • To remain OSHA compliant, your emergency response stations must remain unobstructed and visible.
  • The NoTrax Eyewash Station Mat will ensure that your response station remains visible and unobstructed.
  • This mat is perfect for any emergency response station, including eyewash stations and showers.
  • This emergency mat is highly visible due to its bright safety colors, and its elevated design with beveled ramp edging prevents trips and falls, while the waffle grid design keeps users above hazardous overflow and assures rapid drainage.

Specifications

Antigen CD4
Material PVC
Type Wet Area Mat
Color Green
Certifications/Compliance Conforms to ANSI Z535.1-1998 Standard
Dimensions (L x W) 48 x 54 in.
For Use With (Application) For placement in in front of emergency response stations
Quantity 1 Ea., 25 μg
Surface Perforated Drainage Pattern
Thickness (English) 0.875 in.
Thickness (Metric) 22.225 mm
Length (English) 48 in.
Length (Metric) 1.2192 m
Width (English) 54 in.
Width (Metric) 1.3716 m
Applications Flow Cytometry
Classification Monoclonal
Clone RM4-4
Concentration 0.2 mg/mL
Conjugate APC
Formulation PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
Gene Cd4
Gene Accession No. P06332
Gene Symbols Cd4
Host Species Rat
Purification Method Affinity Chromatography
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12504
Target Species Mouse
Content And Storage 4° C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.