Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Description
- The open space in front of emergency response stations and equipment is often viewed as empty space that can be used for storage
- To remain OSHA compliant, your emergency response stations must remain unobstructed and visible.
- The NoTrax Eyewash Station Mat will ensure that your response station remains visible and unobstructed
- This mat is perfect for any emergency response station, including eyewash stations and showers
- Mat is highly visible due to its bright safety colors, and its elevated design with beveled ramp edging prevents trips and falls, while the waffle grid design keeps users above hazardous overflow and assures rapid drainage
Specifications
Specifications
| Length (Metric) | 1.219 m |
| Type | Wet Area Mat |
| Color | Green |
| Material | PVC |
| For Use With (Application) | For placement in in front of emergency response stations |
| Length (English) | 48 in. |
| Certifications/Compliance | Conforms to ANSI Z535.1-1998 Standard |
| Dimensions (L x W) | 48 x 30 in. |
| Surface | Perforated Drainage Pattern |
| Thickness (English) | 0.875 in. |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?