Learn More
Electron Microscopy Sciences Eyewash with Eye Opener 0.5 OZ
SDP
Catalog No. 501928554
Encompass_Preferred
Multi-purpose first aid eyewash and skin rinse with one "all risks" formula with a four year shelf life. Unique "Eye Opener" eliminates the need for an eye cup and saves precious second in an emergency with it's instant "Twist for Activation" design; very innovatiove and much more effective than traditional eyewash with eyecups. "Eye Opener" allows a "free" hand to open the eye. First ever FDA approved dual claim eyewash to be used as "Eye Wash" as well as "Skin Flush". A Benzalkonium chloride-free formula.
Catalog No. 50-192-8554 Supplier Electron Microscopy Sciences Supplier No. 7059004
Multi-purpose first aid eyewash and skin rinse with one "all risks" formula with a four year shelf life. Unique "Eye Opener" eliminates the need for an eye cup and saves precious second in an emergency with it's instant "Twist for Activation" design; very innovatiove and much more effective than traditional eyewash with eyecups. "Eye Opener" allows a "free" hand to open the eye. First ever FDA approved dual claim eyewash to be used as "Eye Wash" as well as "Skin Flush". A Benzalkonium chloride-free formula.