Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyramex™ Eyewear Display

Catalog No. 19151637 Shop All Pyramex Safety Products Products
Change view
Click to view available options
Color:
White
Device Holds:
36 pieces
Description:
Rotating Display Stand
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Device Holds Description
19-151-637 White 36 pieces Rotating Display Stand
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-151-637 Supplier Pyramex™ Supplier No. DISPLAY

missing translation for 'validMainframeMsg'

Easy to view vertical design

Showcase Pyramex™ eyewear on one of the newly designed displays. Available in a variety of sizes and styles. Pyramex™ displays are offered in Elite, Select and Economical versions, in an easy to view vertical design.

Specifications

Description Rotating Display Stand
Color White
Device Holds 36 pieces
Product Type Eyewear display stand
For Use With (Equipment) For glasses
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.