Learn More
ORS Nasco EYEWEAR WILLSON A700 EA
Catalog No. 502551594
Encompass_Preferred
Honeywell part# A700. Honeywell North A700 Series Safety Glasses Frame Color: Clear Lens Type: Scratch-resistant Lens Tint: Clear Lens Material: Polycarbonate Certifications/Compliance: Meets or exceed ANSI Z87.1-2003 standards. Recommended Applications: Safety
Catalog No. 50-255-1594 Supplier ORS Nasco Supplier No. 763A700
Honeywell part# A700. Honeywell North A700 Series Safety Glasses Frame Color: Clear Lens Type: Scratch-resistant Lens Tint: Clear Lens Material: Polycarbonate Certifications/Compliance: Meets or exceed ANSI Z87.1-2003 standards. Recommended Applications: Safety