Learn More
ORS Nasco EYEWEAR WILLSON A705 EA
Catalog No. 502551593
Encompass_Preferred
Honeywell part# A705. Honeywell North A700 Series Safety Glasses Frame Color: Clear Lens Type: Scratch-resistant Anti-fog Lens Tint: Clear Lens Material: Polycarbonate Certifications/Compliance: Meets or exceed ANSI Z87.1-2003 standards. Recommended Applications: Safety
Catalog No. 50-255-1593 Supplier ORS Nasco Supplier No. 763A705
Honeywell part# A705. Honeywell North A700 Series Safety Glasses Frame Color: Clear Lens Type: Scratch-resistant Anti-fog Lens Tint: Clear Lens Material: Polycarbonate Certifications/Compliance: Meets or exceed ANSI Z87.1-2003 standards. Recommended Applications: Safety