Learn More
Euthanex The EZ-103A Microflex Breather/Nosecone for anestitizing mice and rats
Catalog No. NC9019365
Encompass_Preferred
The EZ-103A Microflex Breather incorporates a patented valve design that mitigates the gas flow to the nosecone mask. Accommodates neonates up to animals weighing 500 grams. Features include low profile that makes the EZ-103A suitable for use under a microscope; vertical and angular adjustments allow precise positioning of animal; tight-fitting nosecone eliminates gas leakage and protects personnel from gas exposure. Comes standard with EZ-110 Mouse Nosecone and Plug, EZ-112 Rat Nosecone and Plug, Plastic Surgery Beds (2).
Catalog No. NC9019365 Supplier Euthanex Supplier No. EZ103A
The EZ-103A Microflex Breather incorporates a patented valve design that mitigates the gas flow to the nosecone mask. Accommodates neonates up to animals weighing 500 grams. Features include low profile that makes the EZ-103A suitable for use under a microscope; vertical and angular adjustments allow precise positioning of animal; tight-fitting nosecone eliminates gas leakage and protects personnel from gas exposure. Comes standard with EZ-110 Mouse Nosecone and Plug, EZ-112 Rat Nosecone and Plug, Plastic Surgery Beds (2).