Learn More
Genevac Inc Genevac EZ-2.4 Elite Evaporator System, Efficient Solvent Removal, Low-Maintenance Dry Scroll Pump, Touchscreen Control Interface
Catalog No. NC3376689
Encompass
The Genevac EZ-2.4 ELITE Evaporator System is a high-performance centrifugal evaporator designed for safe and efficient solvent removal across various life science samples. Its advanced technology enables parallel sample processing while preventing overheating and degradation. The system features a user-friendly touchscreen, an auto-defrost condenser, and an optional Inert Gas Purge, setting it apart as a robust solution for challenging solvents and varied sample formats.
Catalog No. NC3376689 Supplier Genevac Inc Supplier No. 100023738
Add to Cart
Add to Cart
Request a Quote
The Genevac EZ-2.4 ELITE Evaporator System is a high-performance centrifugal evaporator designed for safe and efficient solvent removal across various life science samples. Its advanced technology enables parallel sample processing while preventing overheating and degradation. The system features a user-friendly touchscreen, an auto-defrost condenser, and an optional Inert Gas Purge, setting it apart as a robust solution for challenging solvents and varied sample formats.